EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
Polylactide-based paclitaxel-loaded nanoparticles fabricated by dispersion polymerization: characterization, evaluation in cancer cell lines, and preliminary biodistribution studies.
PMID 24961596
Abstract
The macromonomer method was used to prepare cross-linked, paclitaxel-loaded polylactide (PLA)-polyethylene glycol (stealth) nanoparticles using free-radical dispersion polymerization. The method can facilitate the attachment of other molecules to the nanoparticle surface to make it multifunctional. Proton nuclear magnetic resonance and Fourier transform infrared spectra confirm the synthesis of PLA macromonomer and cross-linking agent. The formation of stealth nanoparticles was confirmed by scanning and transmission electron microscopy. The drug release isotherm of paclitaxel-loaded nanoparticles shows that the encapsulated drug is released over 7 days. In vitro cytotoxicity assay in selected breast and ovarian cancer cell lines reveal that the blank nanoparticle is biocompatible compared with medium-only treated controls. In addition, the paclitaxel-loaded nanoparticles exhibit similar cytotoxicity compared with paclitaxel in solution. Confocal microscopy reveals that the nanoparticles are internalized by MCF-7 breast cancer cells within 1 h. Preliminary biodistribution studies also show nanoparticle accumulation in tumor xenograft model. The nanoparticles are suitable for the controlled delivery of bioactive agents.