EMAIL THIS PAGE TO A FRIEND
The Journal of antimicrobial chemotherapy
Experimental study of the efficacy of daptomycin for the treatment of cephalosporin-resistant pneumococcal meningitis.
PMID 24962033
Abstract
To determine the efficacy of daptomycin for the treatment of penicillin- and cephalosporin-resistant pneumococcal meningitis using in vitro and in vivo methods. In vitro killing curves were determined with clinically achievable CSF antibiotic concentrations. In a rabbit model of pneumococcal meningitis, we studied the efficacy (Δ cfu/mL) of daptomycin used at 15 and 25 mg/kg, comparing it with ceftriaxone 100 mg/kg/24 h and ceftriaxone plus vancomycin 30 mg/kg/24 h over a 26 h period against two different strains: HUB 2349 and ATCC 51916, with MICs of 2 and 32 mg/L of cefotaxime/ceftriaxone, respectively. The penetration of daptomycin into CSF ranged between 9% and 11%. Daptomycin therapy achieved an excellent response, being bactericidal within 2 h of antibiotic administration. Against strain HUB 2349, daptomycin at both doses was as effective as ceftriaxone plus vancomycin. Against the highly resistant strain, daptomycin 25 mg/kg was significantly better than ceftriaxone plus vancomycin at 2 and 6 h. Daptomycin at standard doses, and especially at high doses, may be a useful alternative for the treatment of penicillin- and cephalosporin-resistant pneumococcal meningitis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
76607
Urethane, analytical standard
C3H7NO2