EMAIL THIS PAGE TO A FRIEND
Experimental Biology and Medicine (Maywood, N.J.)
Ethanol extract of propolis protects endothelial cells from oxidized low density lipoprotein-induced injury by inhibiting lectin-like oxidized low density lipoprotein receptor-1-mediated oxidative stress.
PMID 24962173