EMAIL THIS PAGE TO A FRIEND
Journal of Veterinary Science
Intragastric gavage with denatonium benzoate acutely induces neuronal activation in the solitary tract nucleus via the vagal afferent pathway.
PMID 24962407