EMAIL THIS PAGE TO A FRIEND
Oncotarget
High nuclear level of Vav1 is a positive prognostic factor in early invasive breast tumors: a role in modulating genes related to the efficiency of metastatic process.
PMID 24962430