EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
Variable-composition structural optimization and experimental verification of MnB3 and MnB4.
PMID 24962459
Abstract
In combination with variable-composition evolutionary algorithm calculations and first-principles calculations, we have systematically searched for all the stable compounds and their crystal structures in the extensively investigated binary Mn-B system. Our results have uncovered four viable ground-state compounds, with Mn2B, MnB, and MnB4, and previously never reported MnB3 and two metastable compounds, MnB2 and Mn3B4. Our calculations demonstrate that the early characterized mC10 structure of MnB4 showed dynamic instability with large imaginary phonon frequencies and, instead, a new mP20 structure is predicted to be stable both dynamically and thermodynamically, with a considerable energy gain and no imaginary phonon frequencies. The new MnB3 compound crystallizes in the monoclinic mC16 structure which lies 3.2 meV per atom below the MnB (oP8) ↔ MnB4 (mP20) tie-line at T = 0 K. Furthermore, these proposed phases have been verified by our annealed samples after arc-melting synthesis and corresponding powder XRD measurements.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
15580
Boron, ≥95% (boron), amorphous powder
B
266620
Boron, crystalline, 1 cm, 99.7% trace metals basis
B
333360
Boron, crystalline, −60 mesh, 99% trace metals basis
B
GF84487674
Boron, microfoil, disks, 10mm, thinness 0.1μm, specific density 23.5μg/cm2, permanent mylar 3.5μm support, 99.6%
B
GF19976080
Boron, microfoil, disks, 25mm, thinness 0.1μm, specific density 23.5μg/cm2, permanent mylar 3.5μm support, 99.6%
B
GF81914834
Boron, monofilament, 100m, diameter 0.1mm
B
GF54382888
Boron, monofilament, 10m, diameter 0.1mm
B
GF74687080
Boron, monofilament, 10m, diameter 0.2mm
B
GF39354048
Boron, monofilament, 200m, diameter 0.1mm
B
GF97861247
Boron, monofilament, 20m, diameter 0.1mm
B
GF04144586
Boron, monofilament, 20m, diameter 0.2mm
B
GF32655315
Boron, monofilament, 50m, diameter 0.1mm
B
GF70375833
Boron, monofilament, 50m, diameter 0.2mm
B
GF98835069
Boron, monofilament, 5m, diameter 0.1mm
B
GF40338132
Boron, monofilament, 5m, diameter 0.2mm
B
463728
Manganese, powder, ≥99.9% trace metals basis
Mn
266132
Manganese, powder, −325 mesh, ≥99% trace metals basis
Mn
266167
Manganese, chips, thickness <2.0 mm, 99%
Mn
GF83932768
Manganese, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
Mn
GF87355334
Manganese, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
Mn
GF15798730
Manganese, foil, not light tested, 100x100mm, thickness 0.0125mm, permanent polyester support, 98.7%
Mn
GF20192240
Manganese, foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 98.7%
Mn
GF43172210
Manganese, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 98.7%
Mn
GF41027352
Manganese, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 98.7%
Mn
GF81243929
Manganese, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 98.7%
Mn
GF28884549
Manganese, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 98.7%
Mn
GF40287258
Manganese, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 98.7%
Mn
GF94007594
Manganese, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 98.7%
Mn
GF58094635
Manganese, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 98.7%
Mn
GF05008092
Manganese, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 98.7%
Mn