EMAIL THIS PAGE TO A FRIEND
The Journal of Neuroscience
The X-linked mental retardation protein OPHN1 interacts with Homer1b/c to control spine endocytic zone positioning and expression of synaptic potentiation.
PMID 24966368