EMAIL THIS PAGE TO A FRIEND
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Differential effects of intravenous anesthetics on PDGF-BB-induced vascular smooth muscle cell migration.
PMID 24969327
Abstract
Intravenous anesthetics are used during the perioperative and/or postoperative period in critically ill patients. Vascular smooth muscle cells (VSMCs) play important roles in vascular injury repair or restenosis after intervention. We previously reported that platelet-derived growth factor (PDGF)-BB induces VSMC migration via extracellular signal-regulated kinase (ERK) and Akt in a VSMC line, A10 cells. In the present study, we investigated the effects of intravenous anesthetics on PDGF-BB-induced VSMC migration and the mechanism. VSMCs migration was assessed using Boyden chamber, and phosphorylation of each protein kinase was analyzed by Western blotting. Propofol or midazolam but not ketamine or dexmedetomidine suppressed PDGF-BB-induced A10 cells migration in a concentration-dependent manner. The suppressive effects on migration were observed also in human aortic smooth muscle cells. Propofol or midazolam did not affect phosphorylation of PDGF receptor β in A10 cells. Propofol or midazolam failed to affect PDGF-BB-induced phosphorylation of ERK or Akt. On the other hand, propofol or midazolam attenuated PDGF-BB-induced phosphorylation of p38 mitogen-activated protein kinase (MAPK), but did not affect phosphorylation of stress-activated protein kinase/c-Jun N-terminal kinase. Both ketamine and dexmedetomidine had no effect on the phosphorylation of p38 MAPK induced by PDGF-BB. These results strongly suggest that propofol or midazolam inhibits VSMC migration by PDGF-BB via suppression of p38 MAPK activation. Propofol or midazolam may affect VSMC function in critically ill patients.