EMAIL THIS PAGE TO A FRIEND
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Ox-LDL influences peripheral Th17/Treg balance by modulating Treg apoptosis and Th17 proliferation in atherosclerotic cerebral infarction.
PMID 24969587
Abstract
CD4(+)CD25(+) regulatory T (Treg) cells and T-helper 17 (Th17) cells play important roles in acute cerebral infarction (ACI). Our previous findings have suggested that oxidized low-density lipoprotein (Ox-LDL) could influence Treg/Th17 ratio in ACI patients. However, the mechanisms are still not clear. We evaluated the effects of ox-LDL on Th17/Treg cell apoptosis and proliferation in vitro. Our results demonstrated that with increased ox-LDL concentrations, the frequency and suppressive function of Treg cells was decreased while the frequency of Th17 cells was elevated in control subjects. In addition, AnnexinV(+) apoptotic rate, Fas/Fas ligand (FasL) expression, and Caspase-3 activity were escalated in Treg cells while were no significant changes in Th17 cells. Simultaneously, 5-Bromo-29-Deoxyuridine (BrdU) and 3-(4,5-Dimethylthiazol-2-yl)-2,5-Diphenyl Tetrazolium Bromide (MTT) incorporation of Th17 cells was elevated accompanied by upregulated nuclear factor-κB (NF-κB) activity. However, Th17 proliferation was decreased when pre-incubated with Pyrrolidine dithiocarbamate (PDTC, inhibitor of NF-κB activation). Furthermore, there were significant changes induced by ox-LDL in Treg apoptosis, Fas/FasL/Caspase-3 expression of Treg cells, and Th17 proliferation, NF-κB activation of Th17 in ACI patients than in patients with transient ischemic attack (TIA) and control subjects (P<0.01, P<0.05 respectively). These data support that ox-LDL may influence the Th17/Treg balance by modulating Fasmediated Treg apoptosis and NF-κB-associated Th17 proliferation. Ox-LDL also induced a more significant alteration of Treg and Th17 in ACI patients than in TIA and control groups, suggesting a novel role in the pathogenesis of ACI.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300686
Anti-CD3E antibody produced in rabbit, affinity isolated antibody