EMAIL THIS PAGE TO A FRIEND
Biomolecules
Interaction of PTPIP51 with Tubulin, CGI-99 and Nuf2 During Cell Cycle Progression.
PMID 24970130
Abstract
Protein tyrosine phosphatase interacting protein 51 (PTPIP51), also known as regulator of microtubule dynamics protein 3, was identified as an in vitro and in vivo interaction partner of CGI-99 and Nuf-2. PTPIP51 mRNA is expressed in all stages of the cell cycle; it is highly expressed six hours post-nocodazole treatment and minimally expressed one hour post-nocodazole treatment. Recent investigations located PTPIP51 protein at the equatorial plate. This study reports the localization of the PTPIP51/CGI-99 and the PTPIP51/Nuf-2 complex at the equatorial region during mitosis. Moreover, Duolink proximity ligation assays revealed an association of PTPIP51 with the microtubular cytoskeleton and the spindle apparatus. High amounts of phosphorylated PTPIP51 associated with the spindle poles was seen by confocal microscopy. In parallel a strong interaction of PTPIP51 with the epidermal growth factor receptor phosphorylating PTPIP51 at the tyrosine 176 residue was seen. In the M/G1 transition a high level of interaction between PTPIP51 and PTP1B was registered, thus restoring the interaction of PTPIP51 and Raf-1, depleted in mitotic cells. Summarizing these new facts, we conclude that PTPIP51 is necessary for normal mitotic processes, impacting on chromosomal division and control of the MAPK pathway activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
DUO92012
Duolink® In Situ Detection Reagents Brightfield
DUO92013
Duolink® In Situ Detection Reagents FarRed
DUO92014
Duolink® In Situ Detection Reagents Green
DUO92007
Duolink® In Situ Detection Reagents Orange
DUO92008
Duolink® In Situ Detection Reagents Red
DUO92106
Duolink® In Situ Orange Starter Kit Goat/Rabbit
DUO92104
Duolink® In Situ Orange Starter Kit Mouse/Goat
DUO92102
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
DUO92006
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
DUO92003
Duolink® In Situ PLA® Probe Anti-Goat PLUS, Affinity purified Donkey anti-Goat IgG (H+L)
DUO92004
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
DUO92001
Duolink® In Situ PLA® Probe Anti-Mouse PLUS, Affinity purified Donkey anti-Mouse IgG (H+L)
DUO92005
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
DUO92002
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
DUO92010 Duolink® In Situ Probemaker MINUS
DUO92009 Duolink® In Situ Probemaker PLUS
DUO92105
Duolink® In Situ Red Starter Kit Goat/Rabbit
DUO92103
Duolink® In Situ Red Starter Kit Mouse/Goat
DUO92101
Duolink® In Situ Red Starter Kit Mouse/Rabbit