EMAIL THIS PAGE TO A FRIEND
International journal of oncology
Sine oculis homeobox homolog 1 promotes DNA replication and cell proliferation in cervical cancer.
PMID 24970368
Abstract
Malignant proliferation is the fundamental trait of tumor cells. The initiation of DNA replication represents a key process for cell proliferation, and has a marked impact on tumorigenesis and progression. Here we report that Sine oculis homeobox homolog 1 (SIX1) functions as a master regulator in DNA replication of cervical cancer cells. The expression of SIX1 was induced by the E7 oncoprotein of human papillomaviruses in cervical intraepithelial neoplasia and cervical cancer. The increase of SIX1 expression resulted in the upregulation of multiple genes related to the initiation of DNA replication, including the genes coding for the proteins in minichromosome maintenance complex (MCM2, MCM3, MCM6), DNA polymerase α-primase complex (POLA1, PRIM1, PRIM2), clamp loader (RFC3, RFC4, RFC5), DNA polymerase δ complex (POLD3) and DNA polymerase ε complex (POLE2). In line with this, the increase of SIX1 expression enhanced DNA synthesis, accelerated G1 to S phase progression, and promoted the proliferation of cervical cancer cells and the growth of cervical cancer. Consistently, knockdown of SIX1 could hamper DNA synthesis, slow down G1 to S phase progression, and suppress tumor cell proliferation and tumor growth. Importantly, SIX1 could more efficiently promote anchorage-independent cell growth. These results suggest that the increase of SIX1 expression could promote tumorigenesis, progression and invasive growth of cervical cancer by promoting DNA replication, and that targeting SIX1 may have significant therapeutic value in cervical cancer treatment.