EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
HMGB1-DNA complex-induced autophagy limits AIM2 inflammasome activation through RAGE.
PMID 24971542
Abstract
High mobility group box 1 (HMGB1) is a prototype damage-associated molecular pattern (DAMP) that can induce inflammatory and immune responses alone as well as in combination with other molecules such as DNA. However, the intricate molecular mechanisms underlying HMGB1-DNA complex-mediated innate immune response remains largely elusive. In this study, we demonstrated that HMGB1-DNA complex initially induced absent in melanoma 2 (AIM2)-dependent inflammasome activation, and promoted rapid release of inflammasome-dependent early proinflammatory cytokines such as interleukin 1β (IL-1β). Subsequently, HMGB1-DNA complex stimulated an ATG5-dependent cellular degradation process, autophagy, which was paralleled by a cessation of AIM2 inflammasome activation and IL-1β release. These HMGB1-DNA complex-induced inflammasome activation and autophagy were both dependent on the receptor for advanced glycation endproducts (RAGE) that recognizes a wide array of ligands (including HMGB1 and DNA). Thus, autophagy may function as a negative counter-regulatory mechanism for HMGB1-DNA complex-induced inflammasome activation, and provide a checkpoint to limit the development of inflammation.