EMAIL THIS PAGE TO A FRIEND
Virology
Nitazoxanide, an antiviral thiazolide, depletes ATP-sensitive intracellular Ca(2+) stores.
PMID 24971706
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G2752
Gly-Phe
C11H14N2O3
N0290
Nitazoxanide, ≥98% (HPLC)
C12H9N3O5S