EMAIL THIS PAGE TO A FRIEND
Virchows Archiv : an international journal of pathology
Histopathological characterization of ductal carcinoma in situ (DCIS) of the breast according to HER2 amplification status and molecular subtype.
PMID 24973889
Abstract
This study aimed to characterize ductal carcinoma in situ (DCIS) according to human epidermal growth factor receptor 2 (HER2) amplification status and molecular subtype. In addition, we performed a detailed HER2 and CEP17 copy number analysis and we assessed the impact of recent changes in the American Society of Clinical Oncology/College of American Pathologists (ASCO/CAP) guidelines on HER2 immunohistochemical (IHC) scores in DCIS. Nuclear grade, extensive comedonecrosis, stromal architecture, stromal inflammation, and progesterone receptor (PR) expression were significantly associated with HER2 amplification status. In multivariate analysis, stromal inflammation and extensive comedonecrosis were the only two features that remained significantly related to HER2 amplification status. The recent changes in ASCO/CAP guidelines resulted in significant upgrading of HER2 IHC score. Remarkably, about one in five non-amplified DCIS presented a 3+ IHC score, regardless of the scoring method. The biological significance of this phenomenon is presently unknown. After categorization according to molecular subtype, luminal A DCIS mainly presented histopathological features associated with good prognosis, whereas luminal B/HER2+ and HER2+ categories displayed a more aggressive phenotype. Overall, our results demonstrate that HER2-amplified DCIS constitute a clearly distinct subgroup which is characterized by histopathological features associated with poor prognosis. Further studies are required to elucidate the biological significance of a 3+ IHC score in non-amplified DCIS, as well as its mechanism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300344
Anti-ESR1 (Ab-104) antibody produced in rabbit, affinity isolated antibody
SAB4300345
Anti-ESR1 (Ab-106) antibody produced in rabbit, affinity isolated antibody
SAB4300346
Anti-ESR1 (Ab-118) antibody produced in rabbit, affinity isolated antibody
SAB4300347
Anti-ESR1 (Ab-167) antibody produced in rabbit, affinity isolated antibody
SAB2100711
Anti-ESR1 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100712
Anti-ESR1 (ab2) antibody produced in rabbit, affinity isolated antibody
GW21182 Anti-ESR1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA000449
Anti-ESR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA000450
Anti-ESR1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104971
Anti-ESR1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
E0521 Anti-Estrogen Receptor (18-32) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA000451
Anti-MKI67 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA001164
Anti-MKI67 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AMAB90867
Monoclonal Anti-ESR1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1196, purified immunoglobulin, buffered aqueous glycerol solution
SAB1402766
Monoclonal Anti-ESR1 antibody produced in mouse, clone 2F8, purified immunoglobulin, buffered aqueous solution
SAB5300303
Monoclonal Anti-ESR1 antibody produced in mouse, clone 2B7, purified immunoglobulin, buffered aqueous solution
SAB5300304
Monoclonal Anti-ESR1 antibody produced in mouse, clone 5D4B1, ascites fluid
SAB5300305
Monoclonal Anti-ESR1 antibody produced in mouse, clone 10H12B10, ascites fluid
SAB5300306 Monoclonal Anti-ESR1 antibody produced in mouse, clone 8H9A10, ascites fluid
SAB5300307 Monoclonal Anti-ESR1 antibody produced in mouse, clone 2G112B3, ascites fluid
SAB5300308
Monoclonal Anti-ESR1 antibody produced in mouse, clone 6B6, ascites fluid
WH0004288M1
Monoclonal Anti-MKI67 antibody produced in mouse, clone 7B8, purified immunoglobulin, buffered aqueous solution
AMAB90870
Monoclonal Anti-MKI67 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1234, purified immunoglobulin, buffered aqueous glycerol solution
SAB5300423 Monoclonal Anti-MKI67 antibody produced in mouse, clone 4A1, ascites fluid
SAB5300424 Monoclonal Anti-MKI67 antibody produced in mouse, clone 9C12B2, ascites fluid
SAB5300425 Monoclonal Anti-MKI67 antibody produced in mouse, clone 8D5, ascites fluid