EMAIL THIS PAGE TO A FRIEND
Journal of human nutrition and dietetics : the official journal of the British Dietetic Association
Cardiovascular disease biomarkers in patients with inborn errors of protein metabolism: a pilot study.
PMID 24976504
Abstract
Limited data exist so far on cardiovascular disease biomarkers in patients maintained on a protein-restricted diet for inborn errors of protein metabolism. The present study aimed to analyse plasma cholesterol, lipoproteins, triglycerides and total homocysteine in patients with various inborn errors of protein metabolism in comparison with healthy controls. A cross-sectional study of cardiovascular disease biomarkers was conducted in a cohort of patients with inborn errors of protein metabolism: nine phenylketonuria, nine urea cycle defect, six branched chain organic acidaemia and two tyrosinaemia type I patients compared to 30 healthy controls. All patients were on a strict natural protein diet for a mean (SD) period of 5.37 (2.30) years (range 2-9 years). Dietary assessment, plasma cholesterol, triglycerides, lipoproteins and total homocysteine levels were obtained. There were no significant differences in blood lipid studies and total homocysteine levels between patients and controls. The results obtained in this pilot study suggest that cardiovascular disease biomarkers are not increased in patients with inborn errors of protein metabolism. This may be explained by the possible protective effect of a mono- and polyunsaturated fat rich Mediterranean diet. Additional studies with a larger number of patients are needed to confirm this finding.