EMAIL THIS PAGE TO A FRIEND
Brain research
Acute intrastriatal injection of quinolinic acid provokes long-lasting misregulation of the cytoskeleton in the striatum, cerebral cortex and hippocampus of young rats.
PMID 24976581
Abstract
Quinolinic acid (QUIN) is a neuroactive metabolite of the kinurenine pathway, considered to be involved in aging and some neurodegenerative disorders, including Huntington׳s disease. In the present work we have studied the long-lasting effect of acute intrastriatal injection of QUIN (150 nmol/0.5 µL) in 30 day-old rats on the phosphorylating system associated with the astrocytic and neuronal intermediate filament (IF) proteins: glial fibrillary acidic protein (GFAP), and neurofilament (NF) subunits (NFL, NFM and NFH) respectively, until 21 days after injection. The acute administration of QUIN altered the homeostasis of IF phosphorylation in a selective manner, progressing from striatum to cerebral cortex and hippocampus. Twenty four hours after QUIN injection, the IFs were hyperphosphorylated in the striatum. This effect progressed to cerebral cortex causing hypophosphorylation at day 14 and appeared in the hippocampus as hyperphosphorylation at day 21 after QUIN infusion. PKA and PKCaMII have been activated in striatum and hippocampus, since Ser55 and Ser57 in NFL head domain were hyperphosphorylated. However, MAPKs (Erk1/2, JNK and p38MAPK) were hyperphosphorylated/activated only in the hippocampus, suggesting different signaling mechanisms in these two brain structures during the first weeks after QUIN infusion. Also, protein phosphatase 1 (PP1) and 2B (PP2B)-mediated hypophosphorylation of the IF proteins in the cerebral cortex 14 after QUIN injection reinforce the selective signaling mechanisms in different brain structures. Increased GFAP immunocontent in the striatum and cerebral cortex 24h and 14 days after QUIN injection respectively, suggests reactive astrocytes in these brain regions. We propose that disruption of cytoskeletal homeostasis in neural cells takes part of the long-lasting molecular mechanisms of QUIN toxicity in adolescent rats, showing selective and progressive misregulation of the signaling mechanisms targeting the IF proteins in the striatum, cerebral cortex and hippocampus with important implications for brain function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P63204
2,3-Pyridinedicarboxylic acid, 99%
C7H5NO4
HPA041997
Anti-EIF3G antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW22124 Anti-EIF3S4 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405909
Anti-GTF2H4 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV32598
Anti-GTF2H4 antibody produced in rabbit, affinity isolated antibody
SAB4300493
Anti-MAPK3 (Ab-202) antibody produced in rabbit, affinity isolated antibody
SAB4300494
Anti-MAPK3 (Ab-204) antibody produced in rabbit, affinity isolated antibody
SAB1412386
ANTI-MAPK3 antibody produced in mouse, clone 1E2, purified immunoglobulin, buffered aqueous solution
SAB2701851
Anti-mapk3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701770
Anti-mapk3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA003995
Anti-MAPK3 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA005700
Anti-MAPK3 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300497
Anti-MAPK9 (Ab-183) antibody produced in rabbit, affinity isolated antibody
SAB1412229
ANTI-MAPK9 antibody produced in mouse, clone 3C12, purified immunoglobulin, buffered aqueous solution
SAB4502402
Anti-MAPK9 antibody produced in rabbit, affinity isolated antibody
SAB4503423
Anti-MNK1 antibody produced in rabbit, affinity isolated antibody
M3946
Anti-MNK1 antibody produced in rabbit, ~1.4 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4503422
Anti-MNK1 antibody produced in rabbit, affinity isolated antibody
SAB4502389
Anti-p44 MAP Kinase antibody produced in rabbit, affinity isolated antibody
SAB4502390
Anti-p44 MAP Kinase antibody produced in rabbit, affinity isolated antibody
SAB4300200
Anti-phospho-MAPK14 (pThr180) antibody produced in rabbit, affinity isolated antibody
SAB4300201
Anti-phospho-MAPK14 (pTyr182) antibody produced in rabbit, affinity isolated antibody
SAB4300194
Anti-phospho-MAPK3 (pThr202) antibody produced in rabbit, affinity isolated antibody
SAB4300195
Anti-phospho-MAPK3 (pTyr204) antibody produced in rabbit, affinity isolated antibody
SAB4504626
Anti-phospho-MNK1 (pThr255) antibody produced in rabbit, affinity isolated antibody
SAB4504714
Anti-phospho-Mnk1 (pThr385) antibody produced in rabbit, affinity isolated antibody
P1491 Anti-phospho-p38 (pThr180/pTyr182) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4504095
Anti-phospho-p38 MAPK (pThr179+Tyr181) antibody produced in rabbit, affinity isolated antibody
SAB4504497
Anti-phospho-p38 MAPK (pThr180) antibody produced in rabbit, affinity isolated antibody
SAB4504498
Anti-phospho-p38 MAPK (pTyr182) antibody produced in rabbit, affinity isolated antibody