EMAIL THIS PAGE TO A FRIEND
Therapeutic Drug Monitoring
Development and validation of a rapid and simple LC-MS/MS method for quantification of vemurafenib in human plasma: application to a human pharmacokinetic study.
PMID 24977381