EMAIL THIS PAGE TO A FRIEND
Oncotarget
FOXO1 downregulation contributes to the oncogenic program of primary mediastinal B-cell lymphoma.
PMID 24977668
Abstract
Recently we have shown that the transcription factor FOXO1, highly expressed in B cells, is downregulated in classical Hodgkin lymphoma (cHL). As primary mediastinal B cell lymphoma (PMBL) has similarities with the cHL transcription program we investigated FOXO1 expression in this entity. By using immunohistochemistry we found that FOXO1 was absent or expressed at low levels in 19 of 20 primary PMBL cases. PMBL cell lines reproduce the low FOXO1 expression observed in primary cases. By analyzing gene expression profiling data we found that FOXO1 expression inversely correlated with JAK2 in PMBL cases. Targeting JAK2 activity by the small molecular weight inhibitor TG101348 resulted in upregulation of FOXO1 mRNA and protein expression in MedB-1 and U2940 cell lines, and the MYC inhibitor 10058-F4 increased FOXO1 mRNA in MedB-1 cells. Moreover, in MedB-1 cells FOXO1 expression was strongly upregulated by the inhibitor of DNA methylation 5-aza-2-deoxycytidine and by the histone deacetylase inhibitor trichostatin A. Since FOXO1 promoter was unmethylated, this effect is most likely indirect. FOXO1 activation in the FOXO1-negative Med-B1 cell line led to growth arrest and apoptosis, which was accompanied by repression of MYC and BCL2L1/BCLxL. Thus, FOXO1 repression might contribute to the oncogenic program and phenotype of PMBL.