EMAIL THIS PAGE TO A FRIEND
Molecular Pharmaceutics
In vivo evaluation of porous silicon and porous silicon solid lipid nanocomposites for passive targeting and imaging.
PMID 24977922
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M2015
1-Stearoyl-rac-glycerol, ≥99%
C21H42O4
A254
AIDA, solid
C11H11NO4
266620
Boron, crystalline, 1 cm, 99.7% trace metals basis
B
333360
Boron, crystalline, −60 mesh, 99% trace metals basis
B
332445
Boron, amorphous, powder, (submicron)
B
15580
Boron, ≥95% (boron), amorphous powder
B
GF84487674
Boron, microfoil, disks, 10mm, thinness 0.1μm, specific density 23.5μg/cm2, permanent mylar 3.5μm support, 99.6%
B
GF19976080
Boron, microfoil, disks, 25mm, thinness 0.1μm, specific density 23.5μg/cm2, permanent mylar 3.5μm support, 99.6%
B
GF81914834
Boron, monofilament, 100m, diameter 0.1mm
B
GF54382888
Boron, monofilament, 10m, diameter 0.1mm
B
GF74687080
Boron, monofilament, 10m, diameter 0.2mm
B
GF40338132
Boron, monofilament, 5m, diameter 0.2mm
B
GF97861247
Boron, monofilament, 20m, diameter 0.1mm
B
GF04144586
Boron, monofilament, 20m, diameter 0.2mm
B
GF32655315
Boron, monofilament, 50m, diameter 0.1mm
B
GF70375833
Boron, monofilament, 50m, diameter 0.2mm
B
GF98835069
Boron, monofilament, 5m, diameter 0.1mm
B
GF39354048
Boron, monofilament, 200m, diameter 0.1mm
B
746371
Sodium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
INa
793558
Sodium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
INa
439681
Sodium iodide, anhydrous, beads, −10 mesh, 99.999% trace metals basis
INa
409286
Sodium iodide, 99.999% trace metals basis
INa
229911
Sodium iodide, ≥99.99% trace metals basis
INa