EMAIL THIS PAGE TO A FRIEND
Indian Journal of Biochemistry & Biophysics
Anti-proliferative and apoptotic properties of a peptide from the seeds of Polyalthia longifolia against human cancer cell lines.
PMID 24980016