EMAIL THIS PAGE TO A FRIEND
Acta Biomaterialia
Composite pullulan-dextran polysaccharide scaffold with interfacial polyelectrolyte complexation fibers: a platform with enhanced cell interaction and spatial distribution.
PMID 24980061