EMAIL THIS PAGE TO A FRIEND
Nature Reviews. Drug Discovery
Memory: Fingolimod modulates bad memories.
PMID 24981359
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0700 FTY720, ≥98% (HPLC)
C19H33NO2 · HCl