EMAIL THIS PAGE TO A FRIEND
Circulation
Letter by Dumesnil and Pibarot regarding article, "systemic hypertension in low-gradient severe aortic stenosis with preserved ejection fraction".
PMID 24982125