EMAIL THIS PAGE TO A FRIEND
Genome Research
DNA replication and transcription programs respond to the same chromatin cues.
PMID 24985913
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-329
Anti-acetyl-Histone H4 (Lys16) Antibody, clone, Upstate®, from rabbit