EMAIL THIS PAGE TO A FRIEND
Molecular biology reports
NDRG1 expression is related to the progression and prognosis of gastric cancer patients through modulating proliferation, invasion and cell cycle of gastric cancer cells.
PMID 24985974
Abstract
N-myc downstream-regulated gene 1 (NDRG1) has been proposed as a tumor suppressor gene in many different types of tumors, but its potential function and corresponding mechanism are not yet fully elucidated. This study aims to detect the possible function of NDRG1 in gastric cancer progression. In this study, 112 paired gastric cancer tissues and corresponding nonmalignant gastric tissues were utilized to identify the differential protein expression of NDRG1 by immunohistochemistry and its clinical significance was analyzed. Furthermore, 49 of 112 paired gastric specimens were used to detect the differential mRNA expression by real-time PCR. The over expression of NDRG1 in human gastric cancer cell line AGS by PcDNA3.1-NDRG1 transfection was utilized to detect the role of NDRG1 in regulating the biological behavior of gastric cancer. NDRG1 expression was significantly decreased in primary gastric cancer tissues, compared with its corresponding nonmalignant gastric tissues (p < 0.05), and its decreased expression was significantly associated with lymph node metastasis (p < 0.01), invasion depth (p < 0.01) and differentiation (p < 0.05). Additionally, the overall survival rate of gastric cancer patients with high expression of NDRG1 was higher than those with low expression during the follow-up period. NDRG1 overexpression suppressed cells proliferation, invasion and induced a G1 cell cycle arrest in gastric cancer. Furthermore, the down-regulation of NDRG1 in gastric cancer metastatic progression was correlated to E-cadherin and MMP-9. Our results verify that NDRG1 acts as a tumor suppressor gene and may play an important role in the metastasis progression and prognosis of gastric cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW21186 Anti-CDH1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501665
Anti-CDH1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2701861
Anti-cdh1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA004812
Anti-CDH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104222
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
SAB4300693
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
SAB2107370
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
C7855
Anti-Cdh1 antibody,Mouse monoclonal, clone DCS-266, purified from hybridoma cell culture
GW22272A Anti-GC4 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW22272B Anti-GC4 (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
M9570 Anti-Matrix Metalloproteinase-9 antibody produced in goat, affinity isolated antibody
HPA001238
Anti-MMP9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV33090
Anti-MMP9 antibody produced in rabbit, IgG fraction of antiserum
AV47974
Anti-NDRG1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV47975
Anti-NDRG1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2500677
Anti-NDRG1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA006881
Anti-NDRG1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0000999M1
Monoclonal Anti-CDH1 antibody produced in mouse, clone 3F4, purified immunoglobulin, buffered aqueous solution
AMAB90862
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1170, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90863
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1172, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90865
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1180, purified immunoglobulin, buffered aqueous glycerol solution
SAB4700237 Monoclonal Anti-CDH1 antibody produced in mouse, clone 67A4, purified immunoglobulin, buffered aqueous solution
AMAB90805
Monoclonal Anti-MMP9 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0539, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90804
Monoclonal Anti-MMP9 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0538, purified immunoglobulin, buffered aqueous glycerol solution
SAB5300247
Monoclonal Anti-MMP9 antibody produced in mouse, clone 5G3, ascites fluid
SAB5300248
Monoclonal Anti-MMP9 antibody produced in mouse, clone 5C3, ascites fluid
AMAB90806
Monoclonal Anti-MMP9 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0542, purified immunoglobulin, buffered aqueous glycerol solution
SAB1402274
Monoclonal Anti-MMP9 antibody produced in mouse, clone 2H4, purified immunoglobulin, buffered aqueous solution
WH0010397M3
Monoclonal Anti-NDRG1 antibody produced in mouse, clone 2D7, purified immunoglobulin, buffered aqueous solution