EMAIL THIS PAGE TO A FRIEND
Human molecular genetics
CGG repeats in RNA modulate expression of TDP-43 in mouse and fly models of fragile X tremor ataxia syndrome.
PMID 24986919
Abstract
Determining the molecular mechanism(s) leading to Purkinje neuron loss in the neurodegenerative disorder fragile X-associated tremor/ataxia syndrome (FXTAS) is limited by the complex morphology of this cell type. Purkinje neurons are notoriously difficult to isolate and maintain in culture presenting considerable difficultly to identify molecular changes in response to expanded CGG repeat (rCGG)-containing mRNA that induces neurotoxicity in FXTAS. Several studies have uncovered a number of RNA-binding proteins involved in translation that aberrantly interact with the CGG-containing RNA; however, whether these interactions alter the translational profile of cells has not been investigated. Here we employ bacTRAP translational profiling to demonstrate that Purkinje neurons ectopically expressing 90 CGG repeats exhibit a dramatic change in their translational profile even prior to the onset of rCGG-induced phenotypes. This approach identified ∼500 transcripts that are differentially associated with ribosomes in r(CGG)₉₀-expressing mice. Functional annotation cluster analysis revealed broad ontologies enriched in the r(CGG)₉₀ list, including RNA binding and response to stress. Intriguingly, a transcript for the Tardbp gene, implicated in a number of other neurodegenerative disorders, exhibits altered association with ribosomes in the presence of r(CGG)₉₀ repeats. We therefore tested and showed that reduced association of Tardbp mRNA with the ribosomes results in a loss of TDP-43 protein expression in r(CGG)₉₀-expressing Purkinje neurons. Furthermore, we showed that TDP-43 could modulate the rCGG repeat-mediated toxicity in a Drosophila model that we developed previously. These findings together suggest that translational dysregulation may be an underlying mechanism of rCGG-induced neurotoxicity in FXTAS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4500141
Anti-CELF-1 antibody produced in rabbit, affinity isolated antibody
SAB1401532
Anti-CUGBP1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4501766
Anti-Lamin A (Cleaved-Asp230) antibody produced in rabbit, affinity isolated antibody
SAB4501767
Anti-Lamin A (Cleaved-Asp230) antibody produced in rabbit, affinity isolated antibody
SAB4501764
Anti-Lamin A antibody produced in rabbit, affinity isolated antibody
HPA006660
Anti-LMNA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA012010
Anti-PARN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA006314
Anti-PARN antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
440191
Hexamethyldisilazane, reagent grade, ≥99%
C6H19NSi2
379212
Hexamethyldisilazane, ReagentPlus®, 99.9%
C6H19NSi2
52619
Hexamethyldisilazane, for GC derivatization, LiChropur, ≥99.0% (GC)
C6H19NSi2
86944
Hexamethyldisilazane, produced by Wacker Chemie AG, Burghausen, Germany, ≥97.0% (GC)
C6H19NSi2
SAB4100418
Monoclonal Anti-FMR1 antibody produced in mouse, clone FMR1-B317, culture supernatant
SAB1403834
Monoclonal Anti-FMR1 antibody produced in mouse, clone 3E11, purified immunoglobulin, buffered aqueous solution
WH0002332M1
Monoclonal Anti-FMR1 antibody produced in mouse, clone 2D4, purified immunoglobulin, buffered aqueous solution
SAB2103977
Anti-CUGBP1, (N-terminal) antibody produced in rabbit, affinity isolated antibody