EMAIL THIS PAGE TO A FRIEND
The American journal of gastroenterology
IP-10 in pediatric celiac disease and food allergy.
PMID 24989102
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G5004 Gluten from wheat