EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Renal physiology
Mammalian target of rapamycin complex 1 activation in podocytes promotes cellular crescent formation.
PMID 24990893
Abstract
Podocytes play a key role in the formation of cellular crescents in experimental and human diseases. However, the underlying mechanisms for podocytes in promoting crescent formation need further investigation. Here, we demonstrated that mammalian target of rapamycin complex 1 (mTORC1) signaling was remarkably activated and hypoxia-inducible factor (HIF) 1α expression was largely induced in cellular crescents from patients with crescentic glomerular diseases. Specific deletion of Tsc1 in podocytes led to mTORC1 activation in podocytes and kidney dysfunction in mice. Interestingly, 33 of 36 knockouts developed cellular or mixed cellular and fibrous crescents at 7 wk of age (14.19±3.86% of total glomeruli in knockouts vs. 0% in control littermates, n=12-36, P=0.04). All of the seven knockouts developed crescents at 12 wk of age (30.92±11.961% of total glomeruli in knockouts vs. 0% in control littermates, n=4-7, P=0.002). Most notably, bridging cells between the glomerular tuft and the parietal basement membrane as well as the cellular crescents were immunostaining positive for WT1, p-S6, HIF1α, and Cxcr4. Furthermore, continuously administrating rapamycin starting at 7 wk of age for 5 wk abolished crescents as well as the induction of p-S6, HIF1α, and Cxcr4 in the glomeruli from the knockouts. Together, it is concluded that mTORC1 activation in podocytes promotes cellular crescent formation, and targeting this signaling may shed new light on the treatment of patients with crescentic glomerular diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW21075 Anti-CXCR4 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
C3116
Anti-CXCR4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500383
Anti-CXCR4 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1405933
Anti-HIF1A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001275
Anti-HIF1A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104366
Anti-HIF1A antibody produced in rabbit, affinity isolated antibody
SAB2702132
Anti-HIF1A antibody produced in rabbit
SAB4503847
Anti-phospho-4E-BP1 (pSer64) antibody produced in rabbit, affinity isolated antibody
SAB4504328
Anti-phospho-4E-BP1 (pThr36) antibody produced in rabbit, affinity isolated antibody
SAB4504329
Anti-phospho-4E-BP1 (pThr45) antibody produced in rabbit, affinity isolated antibody
SAB4503848
Anti-phospho-4E-BP1 (pThr69) antibody produced in rabbit, affinity isolated antibody
SAB4504686
Anti-phospho-4E-BP1 (pThr70) antibody produced in rabbit, affinity isolated antibody
SAB4300179
Anti-phospho-EIF4EBP1 (pThr45) antibody produced in rabbit, affinity isolated antibody
SAB2102716
Anti-WT1 antibody produced in rabbit, affinity isolated antibody
SAB1411399
Anti-WT1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
PLA0304
Goat anti-HIF1-alpha Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
C6598 Monoclonal Anti-CXCR4 (Fusin) antibody produced in mouse, clone 44716, purified immunoglobulin, lyophilized powder
WH0007852M2
Monoclonal Anti-CXCR4 antibody produced in mouse, clone 1F8, ascites fluid
SAB1404560
Monoclonal Anti-CXCR4, (N-terminal) antibody produced in mouse, clone 2G9, purified immunoglobulin, buffered aqueous solution
SAB5200017
Monoclonal Anti-HIF-1 α antibody produced in mouse, clone ESEE122, 1 mg/mL, purified immunoglobulin
H6536 Monoclonal Anti-HIF-1α antibody produced in mouse, ~1 mg/mL, clone H1α67, purified immunoglobulin, buffered aqueous solution
H6411 Monoclonal Anti-HIF-1α antibody produced in mouse, ~1 mg/mL, clone OZ12, purified immunoglobulin, buffered aqueous solution
WH0007490M1
Monoclonal Anti-WT1 antibody produced in mouse, clone 2H4, purified immunoglobulin, buffered aqueous solution
SAB1404530
Monoclonal Anti-WT1 antibody produced in mouse, clone 3B12, purified immunoglobulin, buffered aqueous solution
SAB1409794
Monoclonal Anti-WT1 antibody produced in mouse, clone 1A12, purified immunoglobulin, buffered aqueous solution
SAB1409795 Monoclonal Anti-WT1 antibody produced in mouse, clone 1E2, purified immunoglobulin, buffered aqueous solution
SAB1409796
Monoclonal Anti-WT1 antibody produced in mouse, clone 1E9, purified immunoglobulin, buffered aqueous solution
PLA0081
Rabbit anti-HIF1-alpha Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.