EMAIL THIS PAGE TO A FRIEND
Autophagy
A phase I/II trial of hydroxychloroquine in conjunction with radiation therapy and concurrent and adjuvant temozolomide in patients with newly diagnosed glioblastoma multiforme.
PMID 24991840