EMAIL THIS PAGE TO A FRIEND
Experimental Cell Research
The formation of cortical actin arrays in human trabecular meshwork cells in response to cytoskeletal disruption.
PMID 24992043
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L5288
Latrunculin B from Latruncula magnifica, ≥80% (HPLC), solid
C20H29NSO5