EMAIL THIS PAGE TO A FRIEND
International Journal of Pharmaceutics
Antitumor activity of PEGylated biodegradable nanoparticles for sustained release of docetaxel in triple-negative breast cancer.
PMID 24992317