EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacology and experimental therapeutics
DOT1L inhibitor EPZ-5676 displays synergistic antiproliferative activity in combination with standard of care drugs and hypomethylating agents in MLL-rearranged leukemia cells.
PMID 24993360
Abstract
EPZ-5676 [(2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-5-((((1r,3S)-3-(2-(5-(tert-butyl)-1H-benzo[d]imidazol-2-yl)ethyl)cyclobutyl)(isopropyl)amino)methyl)tetrahydrofuran-3,4-diol], a small-molecule inhibitor of the protein methyltransferase DOT1L, is currently under clinical investigation for acute leukemias bearing MLL-rearrangements (MLL-r). In this study, we evaluated EPZ-5676 in combination with standard of care (SOC) agents for acute leukemias as well as other chromatin-modifying drugs in cellular assays with three human acute leukemia cell lines: MOLM-13 (MLL-AF9), MV4-11 (MLL-AF4), and SKM-1 (non-MLL-r). Studies were performed to evaluate the antiproliferative effects of EPZ-5676 combinations in a cotreatment model in which the second agent was added simultaneously with EPZ-5676 at the beginning of the assay, or in a pretreatment model in which cells were incubated for several days in the presence of EPZ-5676 prior to the addition of the second agent. EPZ-5676 was found to act synergistically with the acute myeloid leukemia (AML) SOC agents cytarabine or daunorubicin in MOLM-13 and MV4-11 MLL-r cell lines. EPZ-5676 is selective for MLL-r cell lines as demonstrated by its lack of effect either alone or in combination in the nonrearranged SKM-1 cell line. In MLL-r cells, the combination benefit was observed even when EPZ-5676 was washed out prior to the addition of the chemotherapeutic agents, suggesting that EPZ-5676 sets up a durable, altered chromatin state that enhances the chemotherapeutic effects. Our evaluation of EPZ-5676 in conjunction with other chromatin-modifying drugs also revealed a consistent combination benefit, including synergy with DNA hypomethylating agents. These results indicate that EPZ-5676 is highly efficacious as a single agent and synergistically acts with other chemotherapeutics, including AML SOC drugs and DNA hypomethylating agents in MLL-r cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405579
Anti-CD14 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
C3350000
Cytarabine, European Pharmacopoeia (EP) Reference Standard
C9H13N3O5
1162002
Cytarabine, United States Pharmacopeia (USP) Reference Standard
C9H13N3O5
SAB4700103 Monoclonal Anti-CD14 antibody produced in mouse, clone MEM-18, purified immunoglobulin, buffered aqueous solution
SAB4700669 Monoclonal Anti-CD14 antibody produced in mouse, clone B-A8, purified immunoglobulin, buffered aqueous solution
SAB4700097
Monoclonal Anti-CD14 antibody produced in mouse, clone MEM-15, purified immunoglobulin, buffered aqueous solution
C7673
Monoclonal Anti-CD14 antibody produced in mouse, clone UCHM-1, purified immunoglobulin, buffered aqueous solution
AMAB90897
Monoclonal Anti-CD14 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1637, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90898
Monoclonal Anti-CD14 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1638, purified immunoglobulin, buffered aqueous glycerol solution
P6004
Prednisolone, ≥99%
C21H28O5
PHR1043
Prednisolone, Pharmaceutical Secondary Standard; Certified Reference Material
C21H28O5
1555005
Prednisolone, United States Pharmacopeia (USP) Reference Standard
C21H28O5
P2700000
Prednisolone, European Pharmacopoeia (EP) Reference Standard
C21H28O5
46656
Prednisolone, VETRANAL®, analytical standard
C21H28O5
BP464
Prednisolone, British Pharmacopoeia (BP) Assay Standard
C21H28O5
Y0001359
Prednisolone for peak identification, European Pharmacopoeia (EP) Reference Standard
C21H28O5
Y0001355
Prednisolone for system suitability, European Pharmacopoeia (EP) Reference Standard
C21H28O5
B88209
tert-Butyl acetate, ≥99%
C6H12O2
P8511
trans-2-Phenylcyclopropylamine hydrochloride
C9H11N · HCl
P22370
trans-2-Phenylcyclopropylamine hydrochloride, 97%
C9H11N · HCl