EMAIL THIS PAGE TO A FRIEND
Pediatric Research
Exogenous glucagon-like peptide-2 improves outcomes of intestinal adaptation in a distal-intestinal resection neonatal piglet model of short bowel syndrome.
PMID 24995913