EMAIL THIS PAGE TO A FRIEND
FEBS letters
The association of thromboxane A2 receptor with lipid rafts is a determinant for platelet functional responses.
PMID 24996187
Abstract
We have investigated the presence of thromboxane A2 (TXA2) receptor associated with lipid rafts in human platelets and the regulation of platelet function in response to TXA2 receptor agonists when lipid rafts are disrupted by cholesterol extraction. Platelet aggregation with TXA2 analogs U46619 and IBOP was almost blunted in cholesterol-depleted platelets, as well as αIIbβ3 integrin activation and P-selectin exposure. Raft disruption also inhibited TXA2-induced cytosolic calcium increase and nucleotide release, ruling out an implication of P2Y12 receptor. An important proportion of TXA2 receptor (40%) was colocalized at lipid rafts. The presence of the TXA2 receptor associated with lipid rafts in platelets is important for functional platelet responses to TXA2.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0115
PAC-1, ≥98% (HPLC)
C23H28N4O2