EMAIL THIS PAGE TO A FRIEND
Journal of bone and mineral metabolism
Interleukin-1-induced acute bone resorption facilitates the secretion of fibroblast growth factor 23 into the circulation.
PMID 24996526
Abstract
Fibroblast growth factor 23 (FGF23), a central regulator of phosphate and vitamin D metabolism, is mainly produced by osteocytes in bone and exerts its effects on distant organs. Despite its endocrine function, the mechanism controlling serum FGF23 levels is not fully understood. Here we tested the hypothesis that osteoclastic bone resorption may play a role in regulating circulating levels of FGF23, using a mouse model where injections of interleukin (IL)-1β into the subcutaneous tissue over the calvaria induced rapid bone resorption. A significant amount of FGF23 was detected in the extracts from mouse bones, which supports the idea that FGF23 stays in bone for a while after its production. IL-1β-induced bone resorption was associated with elevated serum FGF23 levels, an effect abolished by pre-treatment with pamidronate. Fgf23 expression was not increased in either the calvariae or tibiae of IL-1β-injected mice, which suggests that IL-1β facilitated the entry of FGF23 protein into circulation by accelerating bone resorption rather than increasing its gene expression. The direct effect of IL-1β on bone was confirmed when it increased FGF23 levels in the conditioned media of mouse calvariae in organ culture. Repeated treatment of the cultured calvariae with IL-1β led to a refractory phase, where FGF23 was not mobilized by IL-1β anymore. Consistent with the in vivo results, treatment with IL-1β failed to increase Fgf23 mRNA in isolated primary osteocytes and osteoblasts. These results suggest that FGF23 produced by osteocytes remains in bone, and that rapid bone resorption facilitates its entry into the bloodstream.