EMAIL THIS PAGE TO A FRIEND
European journal of drug metabolism and pharmacokinetics
The synergism mechanism of Rhubarb Anthraquinones on constipation elucidated by comparative pharmacokinetics of Rhubarb extract between normal and diseased rats.
PMID 24996641
Abstract
In the study, it was hypothesized that Rhubarb Anthraquinones synergistically enhanced the purgative effect on constipation rat from the direct and indirect pathway at the same time. A validated HPLC method was successfully applied to elucidate the synergism mechanism from pharmacokinetics aspect after oral administration of Rhubarb extract with a dose of 0.25 g to normal and constipation rats. Comparison of the pharmacokinetic data of normal and constipation rats showed that there were significant differences (p < 0.05) in the main pharmacokinetic parameters. The C max and AUC of emodin in constipation rats were about ten times that of normal rats, while the t 1/2 was remarkably decreased (p < 0.05). However, a significant decrease (p < 0.05) in AUC value for aloe-emodin and rhein was observed in model group compared with normal group. The results may be attributed to the direct action of aloe-emodin and rhein on intestinal cell membranes and the indirect action of emodin on bowel movement through the adjustment by nervous system.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
229075
1,8-Dihydroxy-3-methylanthraquinone, 98%
C15H10O4
17797
Physcion, ≥98.0% (TLC)
C16H12O5