EMAIL THIS PAGE TO A FRIEND
The EMBO journal
NuMA interacts with phosphoinositides and links the mitotic spindle with the plasma membrane.
PMID 24996901
Abstract
The positioning and the elongation of the mitotic spindle must be carefully regulated. In human cells, the evolutionary conserved proteins LGN/Gαi1-3 anchor the coiled-coil protein NuMA and dynein to the cell cortex during metaphase, thus ensuring proper spindle positioning. The mechanisms governing cortical localization of NuMA and dynein during anaphase remain more elusive. Here, we report that LGN/Gαi1-3 are dispensable for NuMA-dependent cortical dynein enrichment during anaphase. We further establish that NuMA is excluded from the equatorial region of the cell cortex in a manner that depends on the centralspindlin components CYK4 and MKLP1. Importantly, we reveal that NuMA can directly associate with PtdInsP (PIP) and PtdInsP2 (PIP2) phosphoinositides in vitro. Furthermore, chemical or enzymatic depletion of PIP/PIP2 prevents NuMA cortical localization during mitosis, and conversely, increasing PIP2 levels augments mitotic cortical NuMA. Overall, our study uncovers a novel function for plasma membrane phospholipids in governing cortical NuMA distribution and thus the proper execution of mitosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA007327
Anti-GPSM2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
HPA034881
Anti-KRT4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410738
Anti-KRT4 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1410850
Anti-NUMA1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2101663 Anti-NUMA1 antibody produced in rabbit, affinity isolated antibody
HPA019841
Anti-NUMA1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA019859
Anti-NUMA1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
HPA043912
Anti-RACGAP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SML0341
Importazole, ≥98% (HPLC)
C20H22N4
EHU129221
MISSION® esiRNA, targeting human GPSM2
EMU051151
MISSION® esiRNA, targeting mouse Gpsm2
37094
Rapamycin, VETRANAL®, analytical standard
C51H79NO13
SAB2105024
Anti-RACGAP1, (N-terminal) antibody produced in rabbit, affinity isolated antibody