EMAIL THIS PAGE TO A FRIEND
International Journal of Pharmaceutics
Combining in vitro and in silico methods for better prediction of surfactant effects on the absorption of poorly water soluble drugs-a fenofibrate case example.
PMID 24997410