EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
A pressurised hot water extraction and liquid chromatography-high resolution mass spectrometry method to determine polar benzotriazole, benzothiazole and benzenesulfonamide derivates in sewage sludge.
PMID 24997513