EMAIL THIS PAGE TO A FRIEND
Clinical & experimental metastasis
Annexin A1 in primary tumors promotes melanoma dissemination.
PMID 24997993
Abstract
Metastatic melanoma is one of the most aggressive forms of skin cancer and has a poor prognosis. We have previously identified Annexin A1 (ANXA1) as a potential murine melanoma-spreading factor that may modulate cell invasion by binding to formyl peptide receptors (FPRs). Here, we report that (1) in a B16Bl6 spontaneous metastasis model, a siRNA-induced decrease in tumoral ANXA1 expression significantly reduced tumoral MMP2 activity and number of lung metastases; (2) in a retrospective study of 61 patients, metastasis-free survival was inversely related to ANXA1 expression levels in primary tumors (HR 3.15 [1.03-9.69], p = 0.045); (3) in human melanoma cell lines, ANXA1 level was positively correlated with in vitro invasion capacity whereas normal melanocytes contained low ANXA1 levels, and (4) the ANXA1 N-terminal peptide ANXA12-26 stimulated MMP2 activity after interaction with FPRs and significantly stimulated the in vitro invasion of melanomas by acting on FPRs. These findings identify ANXA1 as a proinvasive protein in melanoma that holds promise as a potential prognostic marker and therapeutic target.