EMAIL THIS PAGE TO A FRIEND
Biochimica et Biophysica Acta
Ionomycin causes susceptibility to phospholipase A2 while temperature-induced increases in membrane fluidity fail: possible involvement of actin fragmentation.
PMID 24998360
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
323756
Merocyanine 540, Dye content 90 %
C26H32N3NaO6S2
P2141
Phalloidin from Amanita phalloides, ≥90%
C35H48N8O11S