EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences
Identification and elucidation of the structure of in vivo metabolites of diaveridine in chicken.
PMID 24999617