EMAIL THIS PAGE TO A FRIEND
Oncogenesis
ERBB3 is required for metastasis formation of melanoma cells.
PMID 25000258
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0501
SB-590885, ≥98% (HPLC)
C27H27N5O2