EMAIL THIS PAGE TO A FRIEND
Journal of Endourology / Endourological Society
Image-guided robot-assisted prostate ablation using water jet-hydrodissection: initial study of a novel technology for benign prostatic hyperplasia.
PMID 25000418