EMAIL THIS PAGE TO A FRIEND
Chemosphere
Are standard tests sensitive enough to evaluate effects of human pharmaceuticals in aquatic biota? Facing changes in research approaches when performing risk assessment of drugs.
PMID 25000509