EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Particle size distribution and column efficiency. An ongoing debate revived with 1.9μm Titan-C18 particles.
PMID 25001333
Abstract
The mass transfer mechanism in four prototype columns (2.1 and 3.0×50mm, 2.1 and 3.0×100mm) packed with 1.9μm fully porous Titan-C18 particles was investigated by using two previously reported home-made protocols. The first one was used to measure the eddy dispersion HETP of these new columns, the second one to estimate their intrinsic (corrected for HPLC system contribution) HETPs. Titan particles are fully porous particles with a narrow particle size distribution (RSD of 9.2%). The mean Sauter diameter (dSauter=2.04μm) was determined from Coulter counter measurements on the raw silica material (before C18 derivatization) and in the absence of a dispersant agent (Triton X-100) in a 2% NaCl electrolyte solution. The results show that these RPLC Titan columns have intrinsic minimum reduced HETPs ranging from 1.7 to 1.9 and generate up to 290,000 plates per meter. The 3.0mm i.d. columns are more efficient than the 2.1mm i.d. ones and short columns are preferred to minimize efficiency losses due to frictional heating at high speeds. This work also revealed that (1) the lowest h values of the Titan columns are observed at low reduced velocities (νopt=5); (2) this is due to the unusually small diffusivity of analytes across the porous Titan-C18 particles; and (3) the Titan columns are not packed more uniformly than conventional columns packed with fully porous particles. Earlier and recent findings showing that the PSD has no direct physical impact on eddy dispersion and column efficiency are confirmed by these results.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1530
Acetaminophen Related Compound D, Pharmaceutical Secondary Standard; Certified Reference Material
C8H9NO
397237
Acetanilide, zone-refined, purified by sublimation, ≥99.95%
C8H9NO
397229
Acetanilide, purified by sublimation, ≥99.9%
C8H9NO
112933
Acetanilide, 99%
C8H9NO
00401
Acetanilide, puriss. p.a., ≥99.5% (CHN)
C8H9NO
PHR1086
Acetanilide melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
C8H9NO
1004001
Acetanilide Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
C8H9NO
63634
Acetophenone, TraceCERT®, certified reference material
C8H8O
42163
Acetophenone, analytical standard
C8H8O
00790
Acetophenone, puriss. p.a., ≥99.0% (GC)
C8H8O
A10701
Acetophenone, ReagentPlus®, 99%
C8H8O
W200905
Acetophenone, ≥98%, FG
C8H8O
W200910
Acetophenone, natural, 98%, FG
C8H8O
319775
Octanophenone, ≥99%
C14H20O
577120-U Titan C18 UHPLC Column, 1.9 micron, 1.9 μm particle size, L × I.D. 2 cm × 2.1 mm
577121-U Titan C18 UHPLC Column, 1.9 micron, 1.9 μm particle size, L × I.D. 3 cm × 2.1 mm
577122-U Titan C18 UHPLC Column, 1.9 micron, 1.9 μm particle size, L × I.D. 5 cm × 2.1 mm
577123-U Titan C18 UHPLC Column, 1.9 micron, 1.9 μm particle size, L × I.D. 7.5 cm × 2.1 mm
577124-U Titan C18 UHPLC Column, 1.9 micron, 1.9 μm particle size, L × I.D. 10 cm × 2.1 mm
577125-U Titan C18 UHPLC Column, 1.9 micron, 1.9 μm particle size, L × I.D. 3 cm × 3 mm
577126-U Titan C18 UHPLC Column, 1.9 micron, 1.9 μm particle size, L × I.D. 5 cm × 3 mm
577127-U Titan C18 UHPLC Guard Cartridge, 1.9 micron, 1.9 μm particle size, L × I.D. 5 mm × 2.1 mm, pkg of 3 ea
577128-U Titan C18 UHPLC Guard Cartridge, 1.9 micron, 1.9 μm particle size, L × I.D. 5 mm × 3 mm, pkg of 3 ea
V659
Valerophenone, 99%
C11H14O