EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Sorptive extraction using polydimethylsiloxane/metal-organic framework coated stir bars coupled with high performance liquid chromatography-fluorescence detection for the determination of polycyclic aromatic hydrocarbons in environmental water samples.
PMID 25001335
Abstract
In this work, metal-organic frameworks (MOFs, Al-MIL-53-NH₂) were synthesized via the hydrothermal method, and novel polydimethylsiloxane/metal-organic framework (PDMS/MOFs, PDMS/Al-MIL-53-NH₂)-coated stir bars were prepared by the sol-gel technique. The preparation reproducibility of the PDMS/MOFs-coated stir bar was good, with relative standard deviations (RSDs) ranging from 4.8% to 14.9% (n=7) within one batch and from 6.2% to 16.9% (n=6) among different batches. Based on this fact, a new method of PDMS/MOFs-coated stir bar sorptive extraction (SBSE) and ultrasonic-assisted liquid desorption (UALD) coupled with high performance liquid chromatography-fluorescence detection (HPLC-FLD) was developed for the determination of polycyclic aromatic hydrocarbons (PAHs) in environmental water samples. To obtain the best extraction performance for PAHs, several parameters affecting SBSE, such as extraction time, stirring rate, and extraction temperature, were investigated. Under optimal experimental conditions, wide linear ranges and good RSDs (n=7) were obtained. With enrichment factors (EFs) of 16.1- to 88.9-fold (theoretical EF, 142-fold), the limits of detection (LODs, S/N=3) of the developed method for the target PAHs were found to be in the range of 0.05-2.94 ng/L. The developed method was successfully applied to the analysis of PAHs in Yangtze River and East Lake water samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF32887244
Aluminum, wire reel, 10m, diameter 0.05mm, as drawn, 99.99+%
Al
GF94629101
Aluminum, wire reel, 10m, diameter 0.025mm, as drawn, 99.0+%
Al
GF70442209
Aluminum, wire reel, 100m, diameter 1.0mm, as drawn, 99.5%
Al
GF81923238
Aluminum, wire reel, 25m, diameter 0.25mm, as drawn, 99.5%
Al
GF31334093
Aluminum, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.999%
Al
GF22816830
Aluminum, wire reel, 50m, diameter 0.25mm, as drawn, 99.95+%
Al
GF07499010
Aluminum, foam, 150x150mm, thickness 10mm, bulk density 0.2g/cm3, porosity 0.93, aluminum 6101 grade, 98.5%
Al
GF26812043
Aluminum, foam, 150x150mm, thickness 3.2mm, bulk density 0.2g/cm3, porosity 0.93, 16 pores/cm, aluminum 6101 grade, 98.5%
Al
GF24569602
Aluminum, foam, 150x150mm, thickness 30mm, bulk density 0.2g/cm3, porosity 0.93, aluminum 6101 grade, 98.5%
Al
GF33055185
Aluminum, foam, 150x150mm, thickness 6.35mm, bulk density 0.2g/cm3, porosity 0.93, 16 pores/cm, aluminum 6101 grade, 98.5%
Al
GF37306267
Aluminum, foam, 150x150mm, thickness 6.35mm, bulk density 0.2g/cm3, porosity 0.93, 2 pores/cm, aluminum 6101 grade, 98.5%
Al
GF12106897
Aluminum, foam, 150x150mm, thickness 6.35mm, bulk density 0.2g/cm3, porosity 0.93, 4 pores/cm, aluminum 6101 grade, 98.5%
Al
GF10029596
Aluminum, foam, 150x150mm, thickness 6.35mm, bulk density 0.2g/cm3, porosity 0.93, 8 pores/cm, aluminum 6101 grade, 98.5%
Al
GF33610395
Aluminum, foam, 300x400mm, thickness 6.35mm, bulk density 0.2g/cm3, porosity 0.93, aluminum 6101 grade, 98.5%
Al
GF14206047
Aluminum, foam, 33x140mm, thickness 20mm, bulk density 0.2g/cm3, porosity 0.93, aluminum 6101 grade, 98.5%
Al
GF70010222
Aluminum, honeycomb, 150x150mm, thickness 10mm, cell size 3.2mm, cell wall 0.025mm, core density 0g/cm3, phenolic adhesive (resin), 99.5%
Al
GF08071343
Aluminum, honeycomb, 150x150mm, thickness 52.3mm, cell size 6.3mm, cell wall 0.064mm, core density 0.083g/cm3, phenolic adhesive (resin), facing skin aluminum 0.55mm, 99.9%
Al
GF51278070
Aluminum, honeycomb, 300x300mm, thickness 10mm, cell size 3.2mm, cell wall 0.025mm, core density 0g/cm3, phenolic adhesive (resin), 99.5%
Al
GF31987525
Aluminum, honeycomb, 300x300mm, thickness 10mm, cell size 6.3mm, cell wall 0.064mm, core density 0.083g/cm3, phenolic adhesive (resin), 99.5%
Al
GF26773230
Aluminum, honeycomb, 300x300mm, thickness 13.9mm, cell size 6.3mm, cell wall 0.064mm, core density 0.083g/cm3, phenolic adhesive (resin), facing skin aluminum 0.55mm, 99.9%
Al
GF05528718
Aluminum, honeycomb, 300x300mm, thickness 20mm, cell size 13mm, cell wall 0.1mm, core density 0.062g/cm3, phenolic adhesive (resin), 99.5%
Al
GF19881928
Aluminum, honeycomb, 300x300mm, thickness 20mm, cell size 3.2mm, cell wall 0.025mm, core density 0.072g/cm3, phenolic adhesive (resin), 99.5%
Al
GF87997000
Aluminum, honeycomb, 300x300mm, thickness 20mm, cell size 4.8mm, cell wall 0.038mm, core density 0.070g/cm3, phenolic adhesive (resin), 99.5%
Al
GF23840221
Aluminum, honeycomb, 300x300mm, thickness 26.5mm, cell size 6.3mm, cell wall 0.064mm, core density 0.083g/cm3, phenolic adhesive (resin), facing skin aluminum 0.55mm, 99.9%
Al
GF63970498
Aluminum, honeycomb, 300x300mm, thickness 26.5mm, cell size 6.3mm, cell wall 0.064mm, core density 0.083g/cm3, phenolic adhesive (resin), facing skin carbon fiber / epoxy, 99.9%
Al
GF31796725
Aluminum, honeycomb, 300x300mm, thickness 26.5mm, cell size 6.3mm, cell wall 0.064mm, core density 0.083g/cm3, phenolic adhesive (resin), facing skin glass fiber 0.5mm, 99.9%
Al
GF03467202
Aluminum, honeycomb, 300x300mm, thickness 52.3mm, cell size 6.3mm, cell wall 0.064mm, core density 0.083g/cm3, phenolic adhesive (resin), facing skin glass fiber 0.75mm, 99.9%
Al
GF36470178
Aluminum, honeycomb, 300x300mm, thickness 5mm, cell size 13mm, cell wall 0.1mm, core density 0.062g/cm3, phenolic adhesive (resin), 99.5%
Al
GF86118969
Aluminum, honeycomb, 300x300mm, thickness 5mm, cell size 19mm, cell wall 0.064mm, core density 0.029g/cm3, phenolic adhesive (resin), 99.5%
Al
GF04958084
Aluminum, honeycomb, 300x300mm, thickness 5mm, cell size 3.2mm, cell wall 0.025mm, core density 0.072g/cm3, phenolic adhesive (resin), 99.5%
Al