EMAIL THIS PAGE TO A FRIEND
Molecular and Cellular Biology
Nrf2- and ATF4-dependent upregulation of xCT modulates the sensitivity of T24 bladder carcinoma cells to proteasome inhibition.
PMID 25002527
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S0883
Sulfasalazine, analytical standard, ≥98% (HPLC)
C18H14N4O5S