EMAIL THIS PAGE TO A FRIEND
Drug metabolism and disposition: the biological fate of chemicals
Effect of CYP2A6 genetic polymorphism on the metabolic conversion of tegafur to 5-fluorouracil and its enantioselectivity.
PMID 25002745
Abstract
Tegafur (FT), a prodrug of 5-fluorouracil, is a chiral molecule, a racemate of R- and S-isomers, and CYP2A6 plays an important role in the enantioselective metabolism of FT in human liver microsomes (R-FT > S-FT). This study examined the enantioselective metabolism of FT by microsomes prepared from Sf9 cells expressing wild-type CYP2A6 and its variants (CYP2A6*7, *8, *10, and *11) that are highly prevalent in the Asian population. We also investigated the metabolism of coumarin and nicotine, both CYP2A6 probe drugs, in these variants. Enzyme kinetic analyses showed that CYP2A6.7 (I471T) and CYP2A6.10 (I471T and R485L) had markedly lower Vmax values for both enantiomers than wild-type enzyme (CYP2A6.1) and other variant enzymes, whereas Km values were higher in most of the variant enzymes for both enantiomers than CYP2A6.1. The ratios of Vmax and Km values for R-FT to corresponding values for S-FT (R/S ratio) were similar among enzymes, indicating little difference in enantioselectivity among the wild-type and variant enzymes. Similarly, both CYP2A6.7 and CYP2A6.10 had markedly lower Vmax values for coumarin 7-hydroxylase and nicotine C-oxidase activities than CYP2A6.1 and other variant enzymes, whereas Km values were higher in most of the variant enzymes for both activities than CYP2A6.1. In conclusion, the amino acid substitutions in CYP2A6 variants generally resulted in lower affinity for substrates, while Vmax values were selectively reduced in CYP2A6.7 and CYP2A6.10. Consistent R/S ratios among CYP2A6.1 and variant enzymes indicated that the amino acid substitutions had little effect on enantioselectivity in the metabolism of FT.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72609
Coumarin, certified reference material, TraceCERT®
C9H6O2
C4261
Coumarin, ≥99% (HPLC)
C9H6O2
W526509
Coumarin, ≥98%
C9H6O2
01260595
Coumarin, primary reference standard
C9H6O2
Y0000438
Coumarin, European Pharmacopoeia (EP) Reference Standard
C9H6O2
M2670
Magnesium chloride hexahydrate, BioXtra, ≥99.0%
Cl2Mg · 6H2O
M2393
Magnesium chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Cl2Mg · 6H2O
M7304
Magnesium chloride hexahydrate, meets USP testing specifications
Cl2Mg · 6H2O
255777
Magnesium chloride hexahydrate, 99.995% trace metals basis
Cl2Mg · 6H2O
M9272
Magnesium chloride hexahydrate, ACS reagent, 99.0-102.0%
Cl2Mg · 6H2O
M0250
Magnesium chloride hexahydrate, ReagentPlus®, ≥99.0%
Cl2Mg · 6H2O
63068
Magnesium chloride hexahydrate, BioUltra, for molecular biology, ≥99.0% (KT)
Cl2Mg · 6H2O
63064
Magnesium chloride hexahydrate, BioUltra, ≥99.0% (KT)
Cl2Mg · 6H2O
13152
Magnesium chloride hexahydrate, puriss., meets analytical specification of Ph. Eur., BP, FCC, E511, 99-101%, ≤0.0001% Al
Cl2Mg · 6H2O
63033
Magnesium chloride hexahydrate, tested according to Ph Eur
Cl2Mg · 6H2O
93979
Umbelliferone, suitable for fluorescence indicator, ≥98.0% (HPLC)
C9H6O3
H24003
Umbelliferone, 99%
C9H6O3
54826
Umbelliferone, analytical standard
C9H6O3