EMAIL THIS PAGE TO A FRIEND
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine
Silencing survivin activates autophagy as an alternative survival pathway in HCC cells.
PMID 25008566